Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_207677.3 | NP_997560.3 | 2 | 211 | |
GenBank mRNA | AF457576.1 | - | 1 | 76 | |
AF543541.1 | - | 1 | 76 | ||
AK135320.1 | - | 1 | 820 | ||
BC092250.1 | - | 2 | 211 | 5043318 | |
CB589155.1 | - | 176 | |||
CX201206.1 | - | 1 | 63 | ||
CX201977.1 | - | 2 | 208 |