Gene Transcripts

Gene Symbol
B4GALT2
Entrez Gene ID
8704
Gene Name
beta-1,4-galactosyltransferase 2
Gene Aliases
B4Gal-T2, B4Gal-T3, beta4Gal-T2
Chromosome Location
Chr. 1: 43979202 - 43991171 on Build GRCh38
UniGene ID
Hs.632403
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005417.2 NP_001005417.1 7 1144
NM_003780.4 NP_003771.1 7 1344
NM_030587.2 NP_085076.2 7 1145
XM_017002716.1 XP_016858205.1 7 1102
XM_017002717.1 XP_016858206.1 7 1305
GenBank mRNA AB024434.1 - 6 990
AF038660.1 - 6 1034
AK074533.1 - 5 642
AK095873.1 - 7 1772
AK293418.1 - 7 1094
AL137647.1 - 5 539
BC096821.1 - 7 1112 30558762
Y12510.1 - 6 982

View Full Product Details