Gene Transcripts

Gene Symbol
Mgarp
Entrez Gene ID
67749
Gene Name
mitochondria localized glutamic acid rich protein
Gene Aliases
4930583H14Rik, AI195347, CESP-1, HUMMR, Osap, Qsap
Chromosome Location
Chr. 3: 51388412 - 51396547 on Build GRCm38
UniGene ID
Mm.273339
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026358.3 NP_080634.2 3 257
GenBank mRNA AF228502.1 - 3 236
AK006339.1 - 3 219
AK016342.1 - 3 230
AK151401.1 - 3 251
BC012511.1 - 3 250 4500810
BC022740.1 - 3 411 4511440
BC049556.1 - 3 219 6772062
BY328038.1 - 3 257

View Full Product Details