Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024211.3 | NP_077173.1 | 3 | 472 | |
GenBank mRNA | AI931249.1 | - | 3 | 433 | |
AK003957.1 | - | 2 | 111 | ||
AK009487.1 | - | 3 | 418 | ||
AK009824.1 | - | 3 | 372 | ||
AK168951.1 | - | 3 | 397 | ||
AK181540.1 | - | 2 | 154 | ||
AK183372.1 | - | 154 | |||
AK187524.1 | - | 2 | 154 | ||
AK202646.1 | - | 2 | 154 | ||
AK212146.1 | - | 2 | 144 | ||
AK213879.1 | - | 2 | 154 | ||
AK215545.1 | - | 2 | 144 | ||
BC003455.1 | - | 3 | 448 | 2648336 | |
BC019631.1 | - | 3 | 426 | 4023179 |