Gene Transcripts

Gene Symbol
Atad1
Entrez Gene ID
67979
Gene Name
ATPase family, AAA domain containing 1
Gene Aliases
4921525H23Rik, AW107648, Thorase
Chromosome Location
Chr. 19: 32672558 - 32739766 on Build GRCm38
UniGene ID
Mm.27123
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026487.3 NP_080763.2 5 529
GenBank mRNA AK009419.1 - 1 117
AK014967.1 - 5 546
AK030719.1 - 5 549
AK033639.1 - 5 553
AK044925.1 - 5 529
AK075685.1 - 5 574
AK150469.1 - 5 549
AK152059.1 - 5 549
AK165953.1 - 5 564
BC029085.1 - 5 529 3593451
BC043051.1 - 5 514 5708377

View Full Product Details