Gene Transcripts
- Gene Symbol
- 2700060E02Rik
- Entrez Gene ID
- 68045
- Gene Name
- RIKEN cDNA 2700060E02 gene
- Gene Aliases
- Chromosome Location
- Chr. 14: 19811402 - 19823823 on Build GRCm38
- UniGene ID
- Mm.21932
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_026528.3 |
NP_080804.1
|
4, 5 |
460 |
|
GenBank mRNA |
AK003732.1 |
- |
4, 5 |
456 |
|
AK012451.1 |
- |
4, 5 |
424 |
|
AK012460.1 |
- |
4, 5 |
455 |
|
AK012486.1 |
- |
4, 5 |
461 |
|
AK148686.1 |
- |
4, 5 |
402 |
|
AK160555.1 |
- |
4, 5 |
424 |
|
AK160696.1 |
- |
4, 5 |
462 |
|
AK162070.1 |
- |
4, 5 |
412 |
|
AK162433.1 |
- |
4, 5 |
389 |
|
BC023932.1 |
- |
4, 5 |
439 |
|
BC048357.1 |
- |
4, 5 |
455 |
3979120 |
BC096571.1 |
- |
4, 5 |
443 |
963861 |
View Full Product Details