Gene Transcripts

Gene Symbol
2700060E02Rik
Entrez Gene ID
68045
Gene Name
RIKEN cDNA 2700060E02 gene
Gene Aliases
Chromosome Location
Chr. 14: 19811402 - 19823823 on Build GRCm38
UniGene ID
Mm.21932
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026528.3 NP_080804.1 7, 8 677
GenBank mRNA AK003732.1 - 7, 8 673
AK012451.1 - 7, 8 641
AK012460.1 - 7, 8 672
AK012486.1 - 7, 8 678
AK148686.1 - 7, 8 619
AK160555.1 - 7, 8 641
AK160696.1 - 7, 8 679
AK162070.1 - 7, 8 629
AK162433.1 - 7, 8 606
BC023932.1 - 7, 8 655
BC048357.1 - 7, 8 672 3979120
BC096571.1 - 7, 8 660 963861

View Full Product Details