Gene Transcripts

Gene Symbol
Ndufb2
Entrez Gene ID
68198
Gene Name
NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 2
Gene Aliases
1810011O01Rik, 8kDa, AGGG, AI325567, CI-AGGG
Chromosome Location
Chr. 6: 39592551 - 39599472 on Build GRCm38
UniGene ID
Mm.29415
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026612.3 NP_080888.1 2, 3 283
GenBank mRNA AK007495.1 - 2, 3 259
AK010642.1 - 2, 3 290
AK019008.1 - 2, 3 288
AK028334.1 - 292
AK216480.1 - 1, 2 99
BC013510.1 - 2, 3 293 4225177

View Full Product Details