Gene Transcripts

Gene Symbol
Ccdc34
Entrez Gene ID
68201
Gene Name
coiled-coil domain containing 34
Gene Aliases
2810027O19Rik, 4930522P10Rik, AI596202
Chromosome Location
Chr. 2: 110017510 - 110173355 on Build GRCm38
UniGene ID
Mm.181767
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026613.4 NP_080889.3 5 917
GenBank mRNA AK011768.1 - 4 433
AK033555.1 - 5 896
AK140947.1 - 2 2825
AK147056.1 - 5 918
BC049229.1 - 5 650
BC089458.1 - 7 1082
BC108339.1 - 5 901
BC119347.1 - 4 462 8734096
BC120580.1 - 4 462 40129503

View Full Product Details