Gene Transcripts

Gene Symbol
Kdelc2
Entrez Gene ID
68304
Gene Name
KDEL (Lys-Asp-Glu-Leu) containing 2
Gene Aliases
2010004J24Rik, 4833410J10Rik, AW549401
Chromosome Location
Chr. 9: 53383826 - 53401867 on Build GRCm38
UniGene ID
Mm.427519
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_212445.2 NP_997610.1 2 338
GenBank mRNA AK075811.1 - 2 147
BY182320.1 - 2 339

View Full Product Details