Gene Transcripts

Gene Symbol
Tlcd1
Entrez Gene ID
68385
Gene Name
TLC domain containing 1
Gene Aliases
Chromosome Location
Chr. 11: 78176716 - 78180820 on Build GRCm38
UniGene ID
Mm.390375
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291235.1 NP_001278164.1 4 343
NM_001291236.1 NP_001278165.1 4 819
NM_001291237.1 NP_001278166.1 5 527
NM_026708.2 NP_080984.1 4 654
GenBank mRNA AK002265.1 - 4 602
AK046577.1 - 5 2534
AK161721.1 - 554
AK165432.1 - 766
BC005702.1 - 4 546 3598444
BC039182.1 - 4 789 3500908
BG083251.2 - 4 401

View Full Product Details