Gene Transcripts
- Gene Symbol
- Letmd1
- Entrez Gene ID
- 68614
- Gene Name
- LETM1 domain containing 1
- Gene Aliases
- 1110019O13Rik, AI593524, BB130465, BB235638, HCCR-2, HCCR1, MCC-32, Mccr
- Chromosome Location
- Chr. 15: 100468165 - 100479252 on Build GRCm38
- UniGene ID
- Mm.275272
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_134093.2 |
NP_598854.1
|
4, 5 |
556 |
|
GenBank mRNA |
AF287293.1 |
- |
4, 5 |
496 |
|
AF401483.1 |
- |
4, 5 |
389 |
|
AK041312.1 |
- |
3, 4 |
885 |
|
AK045256.1 |
- |
4, 5 |
376 |
|
AK046496.1 |
- |
3, 4 |
360 |
|
AK080221.1 |
- |
4, 5 |
476 |
|
AK154676.1 |
- |
4, 5 |
471 |
|
AK158718.1 |
- |
3, 4 |
333 |
|
AK169341.1 |
- |
4, 5 |
476 |
|
AK185638.1 |
- |
2, 3 |
100 |
|
AK217327.1 |
- |
2, 3 |
100 |
|
BC021361.1 |
- |
1, 2 |
228 |
5010892 |
View Full Product Details