Gene Transcripts

Gene Symbol
Ms4a6d
Entrez Gene ID
68774
Gene Name
membrane-spanning 4-domains, subfamily A, member 6D
Gene Aliases
1110058E16Rik, AI447446, Ms4a11
Chromosome Location
Chr. 19: 11586606 - 11604804 on Build GRCm38
UniGene ID
Mm.290390
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026835.2 NP_081111.1 4 450
GenBank mRNA AF237911.1 - 4 412
AK004295.1 - 4 433
AK152859.1 - 4 430
AK156878.1 - 4 425
AY039037.1 - 4 412
BC018331.1 - 4 387 3964037
BC029738.1 - 4 394 4016611
BG087418.2 - 4 450

View Full Product Details