Gene Transcripts

Gene Symbol
Hemk1
Entrez Gene ID
69536
Gene Name
HemK methyltransferase family member 1
Gene Aliases
2310008M14Rik, AW049265
Chromosome Location
Chr. 9: 107327082 - 107340411 on Build GRCm38
UniGene ID
Mm.259467
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133984.2 NP_598745.2 8 950
GenBank mRNA AK088071.1 - 7 1255
BC011431.1 - 7 895 3981356
BC024428.1 - 7 755 4971671

View Full Product Details