Gene Transcripts

Gene Symbol
Ms4a6b
Entrez Gene ID
69774
Gene Name
membrane-spanning 4-domains, subfamily A, member 6B
Gene Aliases
1810027D10Rik
Chromosome Location
Chr. 19: 11518475 - 11530403 on Build GRCm38
UniGene ID
Mm.436639
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027209.3 NP_081485.2 5 607
GenBank mRNA AF237909.1 - 5 607
AK007613.1 - 538
AK075671.1 - 5 584
AK172345.1 - 681
BC027425.1 - 5 584 3984299

View Full Product Details