Gene Transcripts

Gene Symbol
Zfp715
Entrez Gene ID
69930
Gene Name
zinc finger protein 715
Gene Aliases
2610041B18Rik, mszf15
Chromosome Location
Chr. 7: 43296522 - 43313311 on Build GRCm38
UniGene ID
Mm.45367
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027264.3 NP_081540.3 5 1110
GenBank mRNA BC040201.1 - 6 1283
BC040386.1 - 4 1253 3585171
BC057313.1 - 5 1104 6827768

View Full Product Details