Gene Transcripts

Gene Symbol
Ccdc89
Entrez Gene ID
70054
Gene Name
coiled-coil domain containing 89
Gene Aliases
1700019B01Rik, AU022100, BOIP
Chromosome Location
Chr. 7: 90426312 - 90428664 on Build GRCm38
UniGene ID
Mm.307198
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027298.1 NP_081574.1 1 836
GenBank mRNA AK006105.1 - 1 336
BC060150.1 - 1 571

View Full Product Details