Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004995.3 | NP_004986.1 | 3 | 650 | |
GenBank mRNA | AK291325.1 | - | 3 | 592 | |
AK312657.1 | - | 3 | 598 | ||
BC064803.1 | - | 3 | 584 | 6046773 | |
HE978355.1 | - | 3 | 599 | ||
U41078.1 | - | 3 | 365 | ||
X83535.1 | - | 3 | 484 | ||
X90925.1 | - | 3 | 477 | ||
Z48481.1 | - | 3 | 478 |