Gene Transcripts
- Gene Symbol
- Csnk1g3
- Entrez Gene ID
- 70425
- Gene Name
- casein kinase 1, gamma 3
- Gene Aliases
- 3300002K07Rik, C330049O21Rik
- Chromosome Location
- Chr. 18: 53862082 - 53955684 on Build GRCm38
- UniGene ID
- Mm.368668
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_152809.2 |
NP_690022.2
|
2, 3 |
905 |
|
GenBank mRNA |
CN454971.1 |
- |
1, 2 |
388 |
|
View Full Product Details