Gene Transcripts

Gene Symbol
Fam189a1
Entrez Gene ID
70638
Gene Name
family with sequence similarity 189, member A1
Gene Aliases
5730507A09Rik, AW047451
Chromosome Location
Chr. 7: 64756091 - 65156564 on Build GRCm38
UniGene ID
Mm.441986
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183087.4 NP_898910.2 10 1966
GenBank mRNA AK138167.1 - 8 1462
AK172974.1 - 10 1738
BC049085.1 - 7 1288

View Full Product Details