Gene Transcripts

Gene Symbol
Riok1
Entrez Gene ID
71340
Gene Name
RIO kinase 1 (yeast)
Gene Aliases
3110046C13Rik, 5430416A05Rik, Ad034
Chromosome Location
Chr. 13: 38036989 - 38061433 on Build GRCm38
UniGene ID
Mm.297130
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024242.3 NP_077204.2 4, 5 907
GenBank mRNA AK017312.1 - 4, 5 602
AK152707.1 - 4, 5 656
AK161315.1 - 5, 6 1150

View Full Product Details