Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023626.4 | NP_076115.3 | 5 | 500 | |
GenBank mRNA | AK030364.1 | - | 4 | 307 | |
AK080787.1 | - | 5 | 469 | ||
AK131993.1 | - | 5 | 500 | ||
AK146145.1 | - | 4 | 317 | ||
AY007791.1 | - | 5 | 469 | ||
BC018342.1 | - | 5 | 450 | 4010253 |