Gene Transcripts

Gene Symbol
Syce2
Entrez Gene ID
71846
Gene Name
synaptonemal complex central element protein 2
Gene Aliases
1700013H19Rik, AA407907, Cesc1
Chromosome Location
Chr. 8: 84872111 - 84887446 on Build GRCm38
UniGene ID
Mm.229128
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168244.1 NP_001161716.1 3 810
NM_001168246.1 NP_001161718.1 4 509
NM_027954.3 NP_082230.2 4 491
GenBank mRNA AK005954.1 - 4 492
AK082833.1 - 3 377
AK167332.1 - 3 809
BC094628.1 - 4 386 3374002
BC132534.1 - 4 441 40130811
BY083329.1 - 3 223

View Full Product Details