Gene Transcripts

Gene Symbol
Elp6
Entrez Gene ID
72341
Gene Name
elongator acetyltransferase complex subunit 6
Gene Aliases
2610001P13Rik, 2610002I17Rik, AV159994, Tmem103
Chromosome Location
Chr. 9: 110305192 - 110322102 on Build GRCm38
UniGene ID
Mm.374250
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081381.1 NP_001074850.1 7 841
GenBank mRNA AK011270.1 - 8 997
AK075986.1 - 7 838
BC048732.1 - 7 816 6512688

View Full Product Details