Gene Transcripts

Gene Symbol
Ndufv2
Entrez Gene ID
72900
Gene Name
NADH dehydrogenase (ubiquinone) flavoprotein 2
Gene Aliases
2900010C23Rik
Chromosome Location
Chr. 17: 66078795 - 66101559 on Build GRCm38
UniGene ID
Mm.2206
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278415.1 NP_001265344.1 4, 5 381
NM_028388.3 NP_082664.1 3, 4 284
GenBank mRNA AK078351.1 - 3, 4 228
AK088193.1 - 1 23
AK146998.1 - 3, 4 218
AK150404.1 - 4, 5 318
AK151729.1 - 4, 5 318
AK159460.1 - 3, 4 201
AK166539.1 - 3, 4 245
AK169143.1 - 3, 4 255
BC030946.1 - 3, 4 213 5003524
BY246389.1 - 3, 4 285
CF106105.1 - 2, 3 126

View Full Product Details