Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_201226.1 | NP_957678.1 | 7 | 1612 | |
GenBank mRNA | AK013512.1 | - | 6 | 589 | |
AK129306.1 | - | 7 | 1421 | ||
AK154897.1 | - | 7 | 1642 | ||
AK183492.1 | - | 1 | 25 | ||
AK192094.1 | - | 3 | 232 | ||
BC094573.1 | - | 7 | 1602 | 30354494 | |
BC138462.1 | - | 7 | 1684 | 8861482 | |
BC138463.1 | - | 7 | 1684 | 8861483 |