Gene Transcripts

Gene Symbol
Rassf6
Entrez Gene ID
73246
Gene Name
Ras association (RalGDS/AF-6) domain family member 6
Gene Aliases
1600016B17Rik
Chromosome Location
Chr. 5: 90601875 - 90640737 on Build GRCm38
UniGene ID
Mm.30172
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028478.2 NP_082754.2 8 842
GenBank mRNA AK005472.1 - 8 790
AK132288.1 - 8 804
BC031808.1 - 8 814
BC039934.1 - 8 889

View Full Product Details