Gene Transcripts

Gene Symbol
Mbd3l1
Entrez Gene ID
73503
Gene Name
methyl-CpG binding domain protein 3-like 1
Gene Aliases
1700070G05Rik, 1700095H13Rik, Mbd3l
Chromosome Location
Chr. 9: 18478330 - 18485298 on Build GRCm38
UniGene ID
Mm.160088
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028557.2 NP_082833.1 3 453
GenBank mRNA AK006927.1 - 3 454
AK018931.1 - 3 448
AY038023.1 - 2 402
BC049738.1 - 3 469 6771508
BC061034.1 - 3 459 6707322

View Full Product Details