Gene Transcripts

Gene Symbol
Otud4
Entrez Gene ID
73945
Gene Name
OTU domain containing 4
Gene Aliases
4930431L18Rik, AI449692, D8Ertd69e, mKIAA1046
Chromosome Location
Chr. 8: 79639676 - 79677755 on Build GRCm38
UniGene ID
Mm.34348
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081164.1 NP_001074633.1 2 448
NM_001256033.1 NP_001242962.1 2 448
GenBank mRNA AW494258.1 - 2 448
BC138373.1 - 2 420 8861393
BC145259.1 - 2 420 9053812

View Full Product Details