Gene Transcripts

Gene Symbol
Gtsf1
Entrez Gene ID
74174
Gene Name
gametocyte specific factor 1
Gene Aliases
1700006H03Rik, Cue110
Chromosome Location
Chr. 15: 103402458 - 103430448 on Build GRCm38
UniGene ID
Mm.325325
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028797.1 NP_083073.1 6, 7 499
GenBank mRNA AK005675.1 - 6, 7 477
BU945975.1 - 6, 7 501
CO812705.1 - 6, 7 523

View Full Product Details