Gene Transcripts

Gene Symbol
Mlkl
Entrez Gene ID
74568
Gene Name
mixed lineage kinase domain-like
Gene Aliases
9130019I15Rik
Chromosome Location
Chr. 8: 111311681 - 111338449 on Build GRCm38
UniGene ID
Mm.207971
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029005.2 NP_083281.1 2 243
GenBank mRNA AK018636.1 - 2 519
AK170260.1 - 2 244
BC023755.1 - 2 528 5354568

View Full Product Details