Gene Transcripts

Gene Symbol
Lmbr1l
Entrez Gene ID
74775
Gene Name
limb region 1 like
Gene Aliases
1110013E13Rik, AA415653, D15Ertd735e, Limr
Chromosome Location
Chr. 15: 98903914 - 98918550 on Build GRCm38
UniGene ID
Mm.385020
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029098.3 NP_083374.1 9, 10 942
GenBank mRNA AK003656.1 - 9, 10 874
AK030589.1 - 9, 10 908
AK049110.1 - 9, 10 943
AK143891.1 - 9, 10 903
AK154968.1 - 9, 10 1076
AK170328.1 - 9, 10 903
AK173121.1 - 9, 10 1323
AY052398.1 - 9, 10 785
BC023397.1 - 9, 10 879 5043877

View Full Product Details