Gene Transcripts

Gene Symbol
Usp47
Entrez Gene ID
74996
Gene Name
ubiquitin specific peptidase 47
Gene Aliases
4930502N04Rik, A630020C16Rik
Chromosome Location
Chr. 7: 112022808 - 112111386 on Build GRCm38
UniGene ID
Mm.16974
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133758.3 NP_598519.2 17 2230
NM_177249.3 NP_796223.2 18 2290
GenBank mRNA AK030909.1 - 2 181
AK131138.1 - 8 907
BC024117.1 - 1 56
BC108425.1 - 18 2231 30062566

View Full Product Details