Gene Transcripts

Gene Symbol
NDUFB6
Entrez Gene ID
4712
Gene Name
NADH:ubiquinone oxidoreductase subunit B6
Gene Aliases
B17, CI
Chromosome Location
Chr. 9: 32553526 - 32573184 on Build GRCh38
UniGene ID
Hs.493668
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002493.4 NP_002484.1 2 350
NM_182739.2 NP_877416.1 2 350
GenBank mRNA AF035840.1 - 2 294
AF067167.1 - 2 323
AK289702.1 - 2 304
BC009801.1 - 2 329 4334414
BC016708.1 - 2 364
BC029247.1 - 2 295 5171627
BI669356.1 - 2 329
BP208846.1 - 2 348
CB140469.1 - 2 351
CR456920.1 - 2 226

View Full Product Details