Gene Transcripts

Gene Symbol
Wdr38
Entrez Gene ID
76646
Gene Name
WD repeat domain 38
Gene Aliases
1700123D08Rik
Chromosome Location
Chr. 2: 38998309 - 39001588 on Build GRCm38
UniGene ID
Mm.358777
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029687.3 NP_083963.1 7 805
GenBank mRNA AK007246.1 - 7 796
AK077027.1 - 1053
BC115629.1 - 7 783 40102866

View Full Product Details