Gene Transcripts

Gene Symbol
Cluap1
Entrez Gene ID
76779
Gene Name
clusterin associated protein 1
Gene Aliases
2310030D15Rik, 2610111M03Rik
Chromosome Location
Chr. 16: 3907659 - 3941147 on Build GRCm38
UniGene ID
Mm.458921
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029738.2 NP_084014.1 4 360
GenBank mRNA AK011846.1 - 5 388
AK134648.1 - 4 327
AK166050.1 - 4 441
BC024910.1 - 4 360 3603454

View Full Product Details