Gene Transcripts

Gene Symbol
Arrdc5
Entrez Gene ID
76920
Gene Name
arrestin domain containing 5
Gene Aliases
1700013E09Rik
Chromosome Location
Chr. 17: 56294111 - 56300286 on Build GRCm38
UniGene ID
Mm.329132
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029799.1 NP_084075.1 1 254
GenBank mRNA BC100496.1 - 1 254

View Full Product Details