Gene Transcripts
- Gene Symbol
- Ccdc151
- Entrez Gene ID
- 77609
- Gene Name
- coiled-coil domain containing 151
- Gene Aliases
- AI644415, C330001K17Rik, b2b1885Clo
- Chromosome Location
- Chr. 9: 21989871 - 22002972 on Build GRCm38
- UniGene ID
- Mm.159953
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK031175.1 |
- |
1 |
365 |
|
View Full Product Details