Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163787.1 | NP_001157259.1 | 2, 3 | 517 | |
NM_029939.3 | NP_084215.2 | 2, 3 | 517 | ||
GenBank mRNA | AK021166.1 | - | 2, 3 | 518 | |
AK031175.1 | - | 2, 3 | 518 | ||
BC051179.1 | - | 2, 3 | 436 | ||
BC057069.1 | - | 2, 3 | 420 | 5690884 |