Gene Transcripts

Gene Symbol
Ntng1
Entrez Gene ID
80883
Gene Name
netrin G1
Gene Aliases
A930010C08Rik, AI853992, Lmnt1
Chromosome Location
Chr. 3: 109780050 - 110143472 on Build GRCm38
UniGene ID
Mm.39262
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030699.2 NP_109624.1 6, 7 2358
GenBank mRNA AB038667.1 - 6, 7 2135
AF475069.1 - 5, 6 1263

View Full Product Details