Gene Transcripts

Gene Symbol
NOTCH2
Entrez Gene ID
4853
Gene Name
notch 2
Gene Aliases
AGS2, HJCYS, hN2
Chromosome Location
Chr. 1: 119911553 - 120073449 on Build GRCh38
UniGene ID
Hs.487360
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024408.3 NP_077719.2 31, 32 6086
XM_005270901.3 XP_005270958.1 32, 33 8400
XM_011541519.2 XP_011539821.1 31, 32 11194
XM_011541520.2 XP_011539822.1 31, 32 8104
XM_017001372.1 XP_016856861.1 31, 32 5756
XM_017001373.1 XP_016856862.1 31, 32 5735
GenBank mRNA AF308601.1 - 31, 32 5801
AF315356.1 - 31, 32 5801
AK027869.1 - 5, 6 925

View Full Product Details