Gene Transcripts
- Gene Symbol
- Sytl2
- Entrez Gene ID
- 83671
- Gene Name
- synaptotagmin-like 2
- Gene Aliases
- AI266830, Slp2, mKIAA1597
- Chromosome Location
- Chr. 7: 90302207 - 90410719 on Build GRCm38
- UniGene ID
- Mm.26751
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001040085.2 |
NP_001035174.1
|
14, 15 |
2854 |
|
NM_001040087.2 |
NP_001035176.1
|
5, 6 |
1061 |
|
NM_001040088.2 |
NP_001035177.1
|
5, 6 |
989 |
|
NM_001289583.1 |
NP_001276512.1
|
15, 16 |
2902 |
|
NM_001289584.1 |
NP_001276513.1
|
15, 16 |
2821 |
|
NM_001289585.1 |
NP_001276514.1
|
6, 7 |
1154 |
|
NM_001289586.1 |
NP_001276515.1
|
6, 7 |
1109 |
|
NM_031394.3 |
NP_113571.2
|
14, 15 |
2782 |
|
GenBank mRNA |
AB050742.1 |
- |
5, 6 |
600 |
|
AB057754.1 |
- |
14, 15 |
2241 |
|
AB057755.1 |
- |
8, 9 |
2751 |
|
AB057756.1 |
- |
6, 7 |
1076 |
|
AB057757.1 |
- |
1, 2 |
1067 |
|
AB057760.1 |
- |
14, 15 |
2160 |
|
AB057761.1 |
- |
13, 14 |
2193 |
|
AB057762.1 |
- |
13, 14 |
2121 |
|
AB057763.1 |
- |
2, 3 |
970 |
|
AK027924.1 |
- |
5, 6 |
972 |
|
AK051653.1 |
- |
6, 7 |
1155 |
|
AK173212.1 |
- |
5, 6 |
519 |
|
BC060665.1 |
- |
6, 7 |
2604 |
|
BC138714.1 |
- |
13, 14 |
2092 |
8861736 |
BC145398.1 |
- |
14, 15 |
2212 |
9053956 |
CA559030.1 |
- |
4, 5 |
495 |
|
DV652808.1 |
- |
8, 9 |
806 |
|
View Full Product Details