Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145501.2 | NP_663476.1 | 7 | 1231 | |
GenBank mRNA | AK030725.1 | - | 7 | 1196 | |
AK165136.1 | - | 1 | 1287 | ||
AK185438.1 | - | 3 | 252 | ||
BC089309.1 | - | 6 | 1920 | ||
BC110363.1 | - | 7 | 1193 | 5346917 |