Gene Transcripts

Gene Symbol
C1galt1
Entrez Gene ID
94192
Gene Name
core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase, 1
Gene Aliases
2210410E06Rik, AV284120, T-synthase
Chromosome Location
Chr. 6: 7845224 - 7872042 on Build GRCm38
UniGene ID
Mm.102752
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052993.3 NP_443719.3 2 343
GenBank mRNA AF157962.1 - 2 303
BC064767.1 - 2 234 30077842

View Full Product Details