Gene Transcripts

Gene Symbol
Mael
Entrez Gene ID
98558
Gene Name
maelstrom homolog (Drosophila)
Gene Aliases
4933405K18Rik, AU019877
Chromosome Location
Chr. 1: 166201385 - 166238744 on Build GRCm38
UniGene ID
Mm.26189
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175296.4 NP_780505.1 6, 7 728
GenBank mRNA AK077103.1 - 6, 7 725
AK165801.1 - 6, 7 725
BC050800.1 - 6, 7 728 6744032

View Full Product Details