Gene Transcripts

Gene Symbol
Kcnb2
Entrez Gene ID
98741
Gene Name
potassium voltage gated channel, Shab-related subfamily, member 2
Gene Aliases
9630047L19Rik, BB130875, Kv2.2
Chromosome Location
Chr. 1: 15287383 - 15714214 on Build GRCm38
UniGene ID
Mm.156081
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098528.2 NP_001091998.1 2 2020
GenBank mRNA BC146609.1 - 2 2020

View Full Product Details