Gene Transcripts

Gene Symbol
Sptssa
Entrez Gene ID
104725
Gene Name
serine palmitoyltransferase, small subunit A
Gene Aliases
1110002B05Rik, AA407909, AA409588, AU041967, Ssspta
Chromosome Location
Chr. 12: 54645374 - 54656572 on Build GRCm38
UniGene ID
Mm.485026
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134054.2 NP_598815.2 2 881
GenBank mRNA AK003171.1 - 2 818
AK003275.1 - 2 821
AK003754.1 - 1 277
AK008904.1 - 2 838
AK018380.1 - 2 858
AK137543.1 - 2 856
BC022674.1 - 2 807 4236922

View Full Product Details