Gene Transcripts

Gene Symbol
Fam110c
Entrez Gene ID
104943
Gene Name
family with sequence similarity 110, member C
Gene Aliases
9030611O19Rik, AU019489
Chromosome Location
Chr. 12: 31073968 - 31079940 on Build GRCm38
UniGene ID
Mm.248938
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027828.2 NP_082104.2 1 527
GenBank mRNA AK018545.1 - 1 502
AK139763.1 - 1 635
AK160079.1 - 1 531
BC019466.1 - 1 527 4458596
BC085288.1 - 1 512 5354365

View Full Product Details