Gene Transcripts

Gene Symbol
OPA1
Entrez Gene ID
4976
Gene Name
OPA1, mitochondrial dynamin like GTPase
Gene Aliases
BERHS, MGM1, MTDPS14, NPG, NTG, largeG
Chromosome Location
Chr. 3: 193593144 - 193697811 on Build GRCh38
UniGene ID
Hs.594504
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015560.2 NP_056375.2 22 2436
NM_130831.2 NP_570844.1 21 2328
NM_130832.2 NP_570845.1 22 2382
NM_130833.2 NP_570846.1 22 2439
NM_130834.2 NP_570847.2 23 2490
NM_130835.2 NP_570848.1 23 2493
NM_130836.2 NP_570849.2 23 2547
NM_130837.2 NP_570850.2 24 2601
XM_017006500.1 XP_016861989.1 23 2414
GenBank mRNA AB011139.1 - 22 2257
BC058013.1 - 3 285
BC075805.1 - 22 2356 4214698

View Full Product Details