Gene Transcripts
- Gene Symbol
- Guca1b
- Entrez Gene ID
- 107477
- Gene Name
- guanylate cyclase activator 1B
- Gene Aliases
- BC018258, GCAP2
- Chromosome Location
- Chr. 17: 47384694 - 47392967 on Build GRCm38
- UniGene ID
- Mm.59151
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_146079.1 |
NP_666191.1
|
2, 3 |
452 |
|
GenBank mRNA |
AK033052.1 |
- |
2, 3 |
417 |
|
BC018258.1 |
- |
2, 3 |
380 |
4511650 |
BC018480.1 |
- |
2, 3 |
421 |
4500919 |
BC029017.1 |
- |
2, 3 |
397 |
5359531 |
BC029661.1 |
- |
2, 3 |
407 |
5366068 |
BC031367.1 |
- |
2, 3 |
415 |
5358667 |
BC031916.1 |
- |
2, 3 |
452 |
5369642 |
BC033348.1 |
- |
2, 3 |
402 |
|
BC033358.1 |
- |
2, 3 |
451 |
5368968 |
BC033364.1 |
- |
2, 3 |
402 |
|
BC033380.1 |
- |
2, 3 |
407 |
5366953 |
BC033383.1 |
- |
2, 3 |
408 |
5368064 |
BC033396.1 |
- |
2, 3 |
399 |
|
BC033423.1 |
- |
2, 3 |
404 |
|
BC034331.1 |
- |
2, 3 |
408 |
5357797 |
BC034595.1 |
- |
2, 3 |
407 |
5362191 |
- Gene Symbol
- Actr1b
- Entrez Gene ID
- 226977
- Gene Name
- ARP1 actin-related protein 1B, centractin beta
- Gene Aliases
- 2310066K23Rik, AA960180, AI851923, Arp1b
- Chromosome Location
- Chr. 1: 36698114 - 36710017 on Build GRCm38
- UniGene ID
- Mm.28521
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC027134.1 |
- |
|
371 |
|
View Full Product Details